Learn More
Sigma Aldrich Fine Chemicals Biosciences Antifoam C Emulsion, 500 mL
Catalog No. NC0678813
Encompass
A 30% aqueous emulsion of polydimethylsiloxane concentrate.
Catalog No. NC0678813 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A8011500ML
A 30% aqueous emulsion of polydimethylsiloxane concentrate.