Learn More
Sigma Aldrich Fine Chemicals Biosciences Antifoam SE-15, 100 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A8582100G
Encompass
Aqueous emulsion for bacterial and mammalian systems. Antifoam SE-15 has been used in the preparation of ramified rolling circle amplification (RAM) amplification buffer. It has also been used to find its effects on cell viability, virus infectivity and PCR (polymerase chain reaction) performance.
Catalog No. NC0907256