Learn More
TARGETMOL CHEMICALS INC ANTIFOLATE C2 5MG
SDP
Catalog No. 502580680
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Antifolate C2 (AGF 154) has antitumor activity with an IC of 0.14 nM measured in a growth inhibition assay of human tumor cells.Antifolate C2 can be used for the study of cancer and autoimmune diseases. purity: 98%
Catalog No. 50-258-0680 Supplier TARGETMOL CHEMICALS INC Supplier No. T266365MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Antifolate C2 (AGF 154) has antitumor activity with an IC of 0.14 nM measured in a growth inhibition assay of human tumor cells.Antifolate C2 can be used for the study of cancer and autoimmune diseases. purity: 98%