Learn More
Medchemexpress LLC Antifungal agent 112 | 68380-53-0 | 99.5% | 287.32 | 25 MG
SDP
Catalog No. 5000392990
Encompass_Preferred
Antifungal agent 112 (Compound 1e) exhibits antifungal activity against *Candida albicans*, *Aspergillus niger*, and *Aspergillus fumigatus* with MIC values of 0.0024, 0.0022, and 0.0028 M, respectively. It is for research use only and not sold to patients.
  • Antifungal activity against *Candida albicans*, *Aspergillus niger*, and *Aspergillus fumigatus*.
  • Available in various quantities (5 mg, 10 mg, 25 mg, 50 mg).
  • Controlled substance, not for sale in all territories.
Catalog No. 50-003-92990 Supplier Medchemexpress LLC Supplier No. 25MGHY164620
Antifungal agent 112 (Compound 1e) exhibits antifungal activity against *Candida albicans*, *Aspergillus niger*, and *Aspergillus fumigatus* with MIC values of 0.0024, 0.0022, and 0.0028 M, respectively. It is for research use only and not sold to patients.
  • Antifungal activity against *Candida albicans*, *Aspergillus niger*, and *Aspergillus fumigatus*.
  • Available in various quantities (5 mg, 10 mg, 25 mg, 50 mg).
  • Controlled substance, not for sale in all territories.