Learn More
Medchemexpress LLC Antifungal agent 112 | 68380-53-0 | 99.5% | 287.32 | 5 MG
SDP
Catalog No. 5000391637
Encompass_Preferred
Antifungal agent 112 (Compound 1e) demonstrates antifungal activity against *Candida albicans*, *Aspergillus niger*, and *Aspergillus fumigatus* with MIC values of 0.0024 M, 0.0022 M, and 0.0028 M, respectively. This product is intended for research use only.
  • Exhibits antifungal activity against common fungal pathogens.
  • High purity of 99.50%.
  • Available in various quantities for research needs.
Catalog No. 50-003-91637 Supplier Medchemexpress LLC Supplier No. HY1646205MG
Antifungal agent 112 (Compound 1e) demonstrates antifungal activity against *Candida albicans*, *Aspergillus niger*, and *Aspergillus fumigatus* with MIC values of 0.0024 M, 0.0022 M, and 0.0028 M, respectively. This product is intended for research use only.
  • Exhibits antifungal activity against common fungal pathogens.
  • High purity of 99.50%.
  • Available in various quantities for research needs.