Learn More
Medchemexpress LLC Antifungal agent 49 | 1414861-21-4 | C15H12O4 | 1 MG
SDP
Catalog No. 5000373525
Encompass_Preferred
Antifungal agent 49 is an antifungal compound active against Cryptococcus neoformans. This agent has demonstrated activity against Cryptococcus neoformans and exhibits cytotoxicity in HepDES 19 cells.
  • Active against Cryptococcus neoformans with a MIC value of 49 μM.
  • Exhibits cytotoxicity in HepDES 19 cells with a CC₅₀ of 79 μM.
Catalog No. 50-003-73525 Supplier Medchemexpress LLC Supplier No. HY1534101MG
Antifungal agent 49 is an antifungal compound active against Cryptococcus neoformans. This agent has demonstrated activity against Cryptococcus neoformans and exhibits cytotoxicity in HepDES 19 cells.
  • Active against Cryptococcus neoformans with a MIC value of 49 μM.
  • Exhibits cytotoxicity in HepDES 19 cells with a CC₅₀ of 79 μM.