Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Antifungal agent 98 | 4771-50-0 | 159.18 | 10 G
SDP

Catalog No. 5000446736
Encompass_Preferred

Antifungal agent 98 is a chemical substance for laboratory use and manufacturing, identified by CAS Number 4771-50-0 and a molecular weight of 159.18.

  • Used as a laboratory chemical.
  • Harmful if swallowed.
  • Causes skin irritation.
  • Causes serious eye irritation.
  • May cause respiratory irritation.
  • Stable under recommended storage conditions.
  • Store at 4°C under nitrogen.
  • Solid appearance.
  • Melting point 206-208°C.
  • Boiling point 341°C at 760 mmHg.

Catalog No. 50-004-46736 Supplier Medchemexpress LLC Supplier No. HYW00798010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antifungal agent 98 is a chemical substance for laboratory use and manufacturing, identified by CAS Number 4771-50-0 and a molecular weight of 159.18.

  • Used as a laboratory chemical.
  • Harmful if swallowed.
  • Causes skin irritation.
  • Causes serious eye irritation.
  • May cause respiratory irritation.
  • Stable under recommended storage conditions.
  • Store at 4°C under nitrogen.
  • Solid appearance.
  • Melting point 206-208°C.
  • Boiling point 341°C at 760 mmHg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.