Learn More
Biomatik Corporation Gamma-Aminobutyric Acid (gABA) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12532
Encompass_Preferred
Gamma-Aminobutyric Acid (gABA) Anti-antigen ELISA Kit for detecting gABA in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 3.12-200ng/ml. Sensitivity: 1.18ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Target Synonyms: 4-Aminobutyric Acid. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6695