Learn More
Bioworld Antigen-Antibody Dissociation Kit, 1 Kit
Supplier:  Bioworld 213100021
Encompass_Preferred
Antigen-Antibody Dissociation KitAs part of the Antigen-Antibody dissociation, two reagents are provided to dissociate your antigen-antibody complex from a column either in natural condition of in acidic conditions. For acidic dissociation, a neutralizer is also provided.Dialyze to desalt after dissociating with neutral dissociator. Neutralize your antibody with Neutralizer after elution with acid dissociator.Kit Components:Neutral Dissociator 50mLAcidic Dissociator 50 mLNeutralizer 10mL
Catalog No. 50-198-581