Learn More
Biogenex Laboratories AR Citra Plus Sol. PH-6.2
SDP
Supplier:  Biogenex Laboratories HK08120K
Encompass_Preferred
BioGenex is the inventor of Antigen Retrieval enabling technology. Antigen Retrieval is an effective way of unmasking antigenic epitopes on the surface of formalin-fixed, parraffin-embedded tissue sections using microwave heating. Covered by exclusive patents issued to BioGenex, this method to recover antigenicity in formalin-fixed, paraffin-embedded tissue sections has been routinely practiced in laboratories throughout the world.
Catalog No. NC0359148