Learn More
Biogenex Laboratories 10X ANTIGEN RETREIVAL 100ML
SDP
Catalog No. NC9286844
Encompass_Preferred
Antigen Retrieval Citra Solution,100mL, 10x Concentrated,Unmasks Most Antigens in Formalin-Fixed Tissues,Excellent Results with PR, p53, Ki-67 Antigen
Catalog No. NC9286844 Supplier Biogenex Laboratories Supplier No. HK0865K
Antigen Retrieval Citra Solution,100mL, 10x Concentrated,Unmasks Most Antigens in Formalin-Fixed Tissues,Excellent Results with PR, p53, Ki-67 Antigen