Learn More
Biocare Medical, LLC Antigen Decloaker, 10X
SDP
Supplier:  Biocare Medical, LLC CB910M
Encompass_Preferred
Antigen Decloaker is a citrate buffer heat retrieval solution, pH 6.0. Antigen Decloaker can be used with Biocare's digital electric pressure cooker (Decloaking Chamber), a steamer, a water bath, or a microwave oven. Antigen Decloaker incorporates Assure technology, a color-coded high temperature pH indicator solution. The end user is assured by visual inspection that the solution is at the correct dilution and pH. This product is specially formulated for superior pH stability at high temperatures and will help prevent the possibility of losing pH sensitive antigens. Antigen Decloaker is non-toxic, non-flammable, odorless and sodium azide and thimerosal free.
Catalog No. 50-823-95