Learn More
Abcam Antigen Retrieval Buffer (100 x Tris Buffer, pH 10.0), 250ML
Catalog No. 502649016
Encompass_Preferred
Antigen Retrieval Buffer (100X Tris Buffer, pH 10.0) is a special solution that enables rehydrate and target retrieval in formalin-fixed, paraffin-embedded tissue sections in one step.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-264-9016 Supplier Abcam Supplier No. AB93682250ML
Antigen Retrieval Buffer (100X Tris Buffer, pH 10.0) is a special solution that enables rehydrate and target retrieval in formalin-fixed, paraffin-embedded tissue sections in one step.
The product is subject to the following: Abcam Restricted Use Statement