Learn More
Abcam Antigen Retrieval Buffer (100 x Tris Buffer, pH 10.0), 125ML
Catalog No. NC2152629
Encompass_Preferred
Antigen Retrieval Buffer (100X Tris Buffer, pH 10.0) is a special solution that enables rehydrate and target retrieval in formalin-fixed, paraffin-embedded tissue sections in one step.
The product is subject to the following: Abcam Restricted Use Statement
This item is not returnable. View return policy
Catalog No. NC2152629 Supplier Abcam Supplier No. AB93682125ML
Antigen Retrieval Buffer (100X Tris Buffer, pH 10.0) is a special solution that enables rehydrate and target retrieval in formalin-fixed, paraffin-embedded tissue sections in one step.
The product is subject to the following: Abcam Restricted Use Statement