Learn More
Abcam Antigen Retrieval Buffer (100 x Citrate Buffer), 250ML
Catalog No. NC1841412
Encompass_Preferred
Antigen retrieval 100X citrate buffer (pH 5.1 - 5.5) is a special solution that enables rehydration and antigen retrieval to be performed in formalin-fixed, paraffin-embedded tissue sections with one step. - Cited in over 25 publications - Contains citrate buffer, 1.5% 2-butoxyethanol, 1% hydrochloric acid - Optimized for citrate buffer-treated antibodies - Contains detergent for paraffin emulsification
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1841412 Supplier Abcam Supplier No. AB93678250ML
Antigen retrieval 100X citrate buffer (pH 5.1 - 5.5) is a special solution that enables rehydration and antigen retrieval to be performed in formalin-fixed, paraffin-embedded tissue sections with one step. - Cited in over 25 publications - Contains citrate buffer, 1.5% 2-butoxyethanol, 1% hydrochloric acid - Optimized for citrate buffer-treated antibodies - Contains detergent for paraffin emulsification
The product is subject to the following: Abcam Restricted Use Statement