Learn More
Abcam Antigen Retrieval Buffer (100 x EDTA Buffer, pH 8.0), 125ML
Catalog No. NC0273844
Encompass_Preferred
Antigen Retrieval Buffer (100X EDTA Buffer, pH 8.0) is a special solution that enables rehydrate and target retrieval in formalin-fixed, paraffin-embedded tissue sections in one step.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0273844 Supplier Abcam Supplier No. AB93680125ML
Antigen Retrieval Buffer (100X EDTA Buffer, pH 8.0) is a special solution that enables rehydrate and target retrieval in formalin-fixed, paraffin-embedded tissue sections in one step.
The product is subject to the following: Abcam Restricted Use Statement