Learn More
Abcam Antigen Retrieval Buffer (100 x Tris-EDTA Buffer, pH 9.0), 125ML
Catalog No. NC0273860
Encompass_Preferred
Antigen retrieval 100X tris-EDTA buffer (pH 9.0) enables target retrieval in formalin-fixed, paraffin-embedded tissue sections in one step. - Cited in over 20 publications - Contains 5% tris, 5% tetrasodium ethylene diamine tetraacetate, 1.5% 2-butoxyethanol - Optimized for tris-EDTA buffer (pH 9.0) treated antibodies - Contains detergent for paraffin emulsification
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0273860 Supplier Abcam Supplier No. AB93684125ML
Antigen retrieval 100X tris-EDTA buffer (pH 9.0) enables target retrieval in formalin-fixed, paraffin-embedded tissue sections in one step. - Cited in over 20 publications - Contains 5% tris, 5% tetrasodium ethylene diamine tetraacetate, 1.5% 2-butoxyethanol - Optimized for tris-EDTA buffer (pH 9.0) treated antibodies - Contains detergent for paraffin emulsification
The product is subject to the following: Abcam Restricted Use Statement