Learn More
Vector Laboratories Antigen Unmasking Solution, Tris-Based 250 ml
SDP
Catalog No. NC9800748
Encompass
This Tris-based solution (pH 9.0) is highly effective at revealing antigens in formalin-fixed, paraffin-embedded tissue sections when used with a high temperature treatment procedure. The product is supplied as a convenient 100x concentrated stock, sufficient for 25 L of working solution.
Catalog No. NC9800748 Supplier Vector Laboratories Supplier No. H3301250
This Tris-based solution (pH 9.0) is highly effective at revealing antigens in formalin-fixed, paraffin-embedded tissue sections when used with a high temperature treatment procedure. The product is supplied as a convenient 100x concentrated stock, sufficient for 25 L of working solution.