Learn More
Sino Biological Anti-HA Tag Antibody (HRP), Mouse MAb 100ug
Supplier:  Sino Biological 100028MM10H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, a synthetic peptide corresponding to the HA-tag sequence. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-224-7754