Learn More
American Research Products Inc AntiHuman CD20
Supplier:  American Research Products Inc IVMB052750UG
Encompass
Anti-Human CD20 (Obinutuzumab) - APC Biosimilar is a biosimilar antibody with human reactivity Available in 50ug size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles
Catalog No. 50-003-04418