Learn More
Fluidigm Corp Anti-Human CD223/LAG-3 (11C3C65)-165Ho 25 Tests
SDP
Supplier:  Fluidigm Corp 3165037C
Encompass
Lymphocyte activation gene 3 (LAG-3), also known as CD223, shares homology with CD4, and LAG-3 binding to MHC class II is thought to contribute to the negative regulation of T cell activation.
Catalog No. 50-196-3171