Learn More
Fluidigm Corp Anti-Human CD366/TIM-3 (D5D5R)-154Sm 25 µg
SDP
Supplier:  Fluidigm Corp 3154024D
Encompass
TIM-3 is a type I transmembrane receptor that is constitutively expressed at high levels on NK cells. It is also expressed on specific subsets of CD4+ and CD8+ T cells, on subpopulations of macrophages and DCs, and on monocytes.
Catalog No. 50-196-3478