Learn More
Fluidigm Corp Anti-Human CD8a (D8A8Y)-162Dy 25 µg
SDP
Supplier:  Fluidigm Corp 3162035D
Encompass
CD8 is found on the majority of thymocytes, a subset of peripheral blood T cells, and NK cells. CD8 acts as a co-receptor with MHC class I-restricted T cell receptors in antigen recognition.
Catalog No. 50-196-3488