Learn More
American Research Products Inc AntiHuman IL 12/23
Supplier:  American Research Products Inc IVMB0510100UG
Encompass
Anti-Human IL 12/23 (Briakinumab) - Fc Reduced HRP Biosimilar is a biosimilar antibody with human reactivity Available in 100ug size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles
Catalog No. 50-003-07098