Learn More
Fluidigm Corp Anti-Human/Mouse CD27 (LG.3A10)-171Yb 25 µg
SDP
Supplier:  Fluidigm Corp 3171025D
Encompass
CD27, also known as S152 and T14, is a type 1 membrane protein. It binds to CD70, and plays a key role in regulating B cell activation and immunoglobulin synthesis. It is expressed on a subset of thymocytes, NK cells, some B cells and virtually all mature T cells, and it is upregulated upon T cell activation.
Catalog No. 50-196-3508