Learn More
American Research Products Inc AntiHuman TIGIT
Supplier:  American Research Products Inc IVMB0267100MG
Encompass
Anti-Human TIGIT [4E1 2] In Vivo Antibody - Low Endotoxin is a in vivo antibody with human reactivity produced in mouse Available in 100 mg size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles
Catalog No. 50-003-06715
Only null left
Add to Cart
Request a Quote