Learn More
Medchemexpress LLC Antimalarial agent 1 | 84176-65-8 | 98.57% | 344.31 | 50 MG
SDP
Catalog No. 5000367038
Encompass_Preferred
Antimalarial agent 1 is a potent antimalarial agent. It is intended for research use only.
  • Potent antimalarial agent.
  • Antimalarial index of 1:40 when tested against Plasmodium gallinaceum.
Catalog No. 50-003-67038 Supplier Medchemexpress LLC Supplier No. HYW00910950MG
Antimalarial agent 1 is a potent antimalarial agent. It is intended for research use only.
  • Potent antimalarial agent.
  • Antimalarial index of 1:40 when tested against Plasmodium gallinaceum.