Learn More
TARGETMOL CHEMICALS INC Antimalarial agent 30 1MG
SDP
Catalog No. 502876784
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Antimalarial agent 30 has anti-Plasmodium berghei liver stage parasite activity and antimalarial activity for the study of malarial infections. Purity 99.94%
Catalog No. 50-287-6784 Supplier TARGETMOL CHEMICALS INC Supplier No. T795351MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Antimalarial agent 30 has anti-Plasmodium berghei liver stage parasite activity and antimalarial activity for the study of malarial infections. Purity 99.94%