Learn More
Med Vet International Norbrook Enroflox 100 (Enrofloxacin), Antimicrobial Injectable Solution, 250mL, Each
Catalog No. 502827762
Encompass_Preferred
Cattle - Single-Dose Therapy: Enroflox 100 (enrofloxacin) is indicated for the treatment of bovine respiratory disease (BRD) associated with Mannheimia haemolytica, Pasteurella multocida, Histophilus somni and Mycoplasma bovis in beef and non-lactating dairy cattle; and for the control of BRD in beef and non-lactating dairy cattle at high risk of developing BRD associated with M. haemolytica, P. multocida, H. somni and M. bovis. Cattle - Multiple-Day Therapy: Enroflox 100 (enrofloxacin) is indicated for the treatment of bovine respiratory disease (BRD)Swine: Enroflox 100 (enrofloxacin) is indicated for the treatment and control of swine respiratory disease (SRD) associated with Actinobacillus pleuropneumoniae, Pasteurella multocida, Haemophilus parasuis, Streptococcus suis, Bordetella bronchiseptica and Mycoplasma hyopneumoniae. Enroflox 100 is indicated for the control of colibacillosis in groups or pens of weaned pigs
Catalog No. 50-282-7762 Supplier Med Vet International Supplier No. RXNOR250INJV
Cattle - Single-Dose Therapy: Enroflox 100 (enrofloxacin) is indicated for the treatment of bovine respiratory disease (BRD) associated with Mannheimia haemolytica, Pasteurella multocida, Histophilus somni and Mycoplasma bovis in beef and non-lactating dairy cattle; and for the control of BRD in beef and non-lactating dairy cattle at high risk of developing BRD associated with M. haemolytica, P. multocida, H. somni and M. bovis. Cattle - Multiple-Day Therapy: Enroflox 100 (enrofloxacin) is indicated for the treatment of bovine respiratory disease (BRD)Swine: Enroflox 100 (enrofloxacin) is indicated for the treatment and control of swine respiratory disease (SRD) associated with Actinobacillus pleuropneumoniae, Pasteurella multocida, Haemophilus parasuis, Streptococcus suis, Bordetella bronchiseptica and Mycoplasma hyopneumoniae. Enroflox 100 is indicated for the control of colibacillosis in groups or pens of weaned pigs