Learn More
Marketlab Antimicrobial Biohazard Shield
Description
- Shield protects from accidental, dangerous spills and splashes
- Shield itself is protected from microorganism growth with AMGARD™ silver ion antimicrobial technology
- Silver ion technology is built into the acrylic, not applied on top, for longer-lasting protection
- Helps reduce degradation of the shield, provides additional surface protection between cleanings, and is active for life of the product
- 2 sizes available
- Mold, mildew, and bacteria can stain your shield and impede your view
- To prevent that from happening, you need an Antimicrobial Biohazard Shield from Marketlab
- These shields are made of clear, scratch-resistant, and chemical-resistant acrylic that's formulated with AMGARD™ silver ion antimicrobial technology
- It's proven to reduce the growth of microorganisms on plastics. And because the silver ion is built in and not sprayed on, it won't wear off
- Your shield is protected for life
Specifications
Specifications
| Lens Coating | Uncoated |
| Height (English) | 15 in. |
| Width (English) | 11.5 in. |
| Depth (English) | 7.5 in. |
| Product Type | Countertop Shield |
| Size | Small |
| Material | Acrylic |
| Disposable | Reusable |
| Lens Material | None |
| UV Protection | None |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.