Learn More
BEST SANITIZERS SoftenSure Foam is a foaming soap that enhances the skin's protective barrier while cleansing. Available in 500mL pump bottle, 1.2L cartridge. fits BSI manual and touchless dispensers
Supplier:  BEST SANITIZERS BHC2006
Encompass_Preferred
SoftenSure Foam is an ideal foaming soap. Engineered with a powerful Benzalkonium Chloride formula, SoftenSure enhances the skin's protective barrier while cleansing at the same time. 0.12% Benzalkonium Chloride (BAC) formula, pH balanced and formulated with emollients to keep skin healthy. NSF certified, E2 classification. A Rich, thick foam that is fragrance and dye free. Kosher, Pareve and Halal certified. Available in 500mL pump bottle, and a 1.2L cartridge. Dispensing options: BSI manual and touchless dispensers 4/CSHand wash
Catalog No. NC3338776