Learn More
BEST SANITIZERS SoftenSure Foam is a foaming soap that enhances the skin's protective barrier while cleansing. Available in 500mL pump bottle, 1.2L cartridge. fits BSI manual and touchless dispensers
Catalog No. NC3338774
Encompass_Preferred
SoftenSure Foam is an ideal foaming soap. Engineered with a powerful Benzalkonium Chloride formula, SoftenSure enhances the skin's protective barrier while cleansing at the same time. 0.12% Benzalkonium Chloride (BAC) formula, pH balanced and formulated with emollients to keep skin healthy. NSF certified, E2 classification. A Rich, thick foam that is fragrance and dye free. Kosher, Pareve and Halal certified. Available in 500mL pump bottle, and a 1.2L cartridge. Dispensing options: BSI manual and touchless dispensers 12/CSHand wash
Catalog No. NC3338774 Supplier BEST SANITIZERS Supplier No. BHC2005
SoftenSure Foam is an ideal foaming soap. Engineered with a powerful Benzalkonium Chloride formula, SoftenSure enhances the skin's protective barrier while cleansing at the same time. 0.12% Benzalkonium Chloride (BAC) formula, pH balanced and formulated with emollients to keep skin healthy. NSF certified, E2 classification. A Rich, thick foam that is fragrance and dye free. Kosher, Pareve and Halal certified. Available in 500mL pump bottle, and a 1.2L cartridge. Dispensing options: BSI manual and touchless dispensers 12/CSHand wash