Learn More
PURELL™ Antimicrobial Foaming Hand Soap
GREENER_CHOICESDP
Catalog No. 19043361 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Solution Type:
Fragrance-free
Light Scent
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Solution Type
19-043-361 Light Scent
19-043-360 Fragrance-free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-043-361 Supplier PURELL™ Supplier No. 838202
Add to Cart
Add to Cart
Refill (1200 mL) for PURELL™ ES10 touch-free hand sanitizer dispensers. Available in light scent or fragrance-free.
  • Effectively kill germs and care for hands, even with frequent use. PURELL™ Antimicrobial Foaming Hand Soap is a quick-acting antimicrobial foam soap for general handwashing that helps reduce germs on skin that could cause disease. This specially formulated refreshing foam helps maintain the skin's natural moisture barrier. Hypoallergenic, it is proven to be as mild as our top-selling non-antimicrobial soap. It is free of dyes, phthalates, parabens, and triclosan, and is a USDA Certified Biobased Product.
    This 1200 mL refill bottle is made with recyclable plastic components. The collar and alkaline AA battery easily pull apart making it easy to recycle and reduce product waste.
    Case Includes: 2 - PURELL™ Antimicrobial Foaming Hand Soap 1200 mL Refills for the ES10 Automatic Soap Dispenser. ES10 dispensers (SKUs 8334-E1, 8330-E1, 8338-E1) are sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hand Sanitizer
Color Light Blue
Form Foam
Size 1200mL
Solution Type Light Scent
Volume (Metric) 1200 mL