Learn More
PURELL™ Antimicrobial Fragrance-Free Foaming Hand Soap PROMO
SDP
Catalog No. 19043360W25 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
PURELL™ Antimicrobial Fragrance-Free Soap
Catalog No.
19-043-360W25 PURELL™ Antimicrobial Fragrance-Free Soap
1 options
Catalog No. 19-043-360W25 Supplier PURELL™ Supplier No. PI838302
Refill (1200 mL) for PURELL™ ES10 touch-free hand sanitizer dispensers.
  • PURELL™ Antimicrobial Foaming Hand Soap is a one-step handwash that provides quick, easy cleaning in food-contact and healthcare settings. Listed with NSF International as E2 for use in all departments of USDA-regulated food-processing environments. Removes fats, oils, and other food-processing soils. USDA Certified Biobased Product. Certified Kosher. Exceptionally mild for frequent use. Hypoallergenic. Free of fragrance, dyes, phthalates, parabens, and triclosan.
    This 1200 mL refill bottle is made with recyclable plastic components. The collar and alkaline AA battery easily pull apart making it easy to recycle and reduce product waste.
    Case Includes: 2 - PURELL™ Antimicrobial Foaming Hand Soap, Fragrance Free, 1200 mL Refills for the ES10 Automatic Soap Dispenser. ES10 dispensers (SKUs 8334-E1, 8330-E1, 8338-E1) are sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hand Sanitizer
Color Light Blue
Form Foam
Size 1200 mL