Learn More
BEST SANITIZERS Softensure Antimicrobial Liquid Soap is 99.999% Effective againgst 29 pathogens in 30 seconds. Gentle on hands and skin even with repeated use. Available in a 500ml pump bottle and a 1.2L cartridge.
Catalog No. NC3338778
Encompass_Preferred
Softensure Antimicrobial Liquid Soap is engineered as a powerful chloroxylenol (PCMX) formula that is 99.999% Effective againgst 29 pathogens in 30 seconds. Formulated with emollients to help keep skin soft and healthy. Rich, thick lather that Enhances the skin's protective barrier while cleansing at the same time. Gentle on hands and skin even with repeated use. Available in a 500ml pump bottle and a 1.2L cartridge. 12/CS
Catalog No. NC3338778 Supplier BEST SANITIZERS Supplier No. BHC2010
Add to Cart
Add to Cart
Softensure Antimicrobial Liquid Soap is engineered as a powerful chloroxylenol (PCMX) formula that is 99.999% Effective againgst 29 pathogens in 30 seconds. Formulated with emollients to help keep skin soft and healthy. Rich, thick lather that Enhances the skin's protective barrier while cleansing at the same time. Gentle on hands and skin even with repeated use. Available in a 500ml pump bottle and a 1.2L cartridge. 12/CS