Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Antimicrobial Properties Of Essential Oils
SCI_ED

Catalog No. S01975 Shop All Aldon Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S01975 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S01975 Supplier Innovating Science™ Supplier No. IS3055
Add to Cart
Add to Cart

Essential oils are commonly used in cleaning products, but which ones are most effective? In this activity, students will perform the Kirby-Bauer test on a variety of essential oils to determine their antimicrobial efficacy.

Specifications

Grade 6+
Class Size 15 Lab Groups
Product Type Chemistry Education Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.