Learn More
Marketlab Antimicrobial Splash Guard with Cutaway
Catalog No. 22505169
Change view
Click to view available options
Size:
Medium
Catalog No. Size
22-505-169 Medium
1 options
Catalog No. 22-505-169 Supplier Marketlab Supplier No. 107393
Add to Cart
Add to Cart
Medium 4.5 W x 23.625 L x 18 H in.
  • Splashes and Germs Have Met Their Match
  • AMGARD™ transparent acrylic inhibits microbe surface degradation
  • Ideal for creating a barrier between two people
  • 4.5 in. wide split-design base for optimum stability
  • Portable design is easy to move to provide protection where you need it most
  • Helps users remain compliant with regulations prohibiting patient care items from entering splash zones
  • Transparent acrylic material inhibits microbe surface degradation for durability and longevity
Specifications
Lens Coating Uncoated
Lens Tint None
Product Type Single Splash Shield
Size Medium
Material Acrylic
Disposable Reusable
Lens Material None
UV Protection None