Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony ICP Standard, 1000 ppm Sb in H2O/Tartaric Acid/tr HNO3, Ricca Chemical
SDP

Catalog No. PSB1KW500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PSB1KW-500 500 mL Natural Poly Bottle
PSB1KW-100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PSB1KW-500 Supplier Ricca Chemical Company Supplier No. PSB1KW500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7440-36-0 , 7697-37-2 , 7647-01-0 , 87-69-4 , 7732-18-5
Molecular Formula Sb
Molecular Weight (g/mol) 121.76
MDL Number MFCD00011211 MFCD00134030
InChI Key WATWJIUSRGPENY-UHFFFAOYSA-N
PubChem CID 5354495
ChEBI CHEBI:30304
IUPAC Name antimony
SMILES [Sb]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 1,000ppm Sb
CAS Min % 96.0, 0.72, 0.57, 0.14, 0.1
CAS Max % 100.0, 0.75, 0.59, 0.15, 0.1
Equivalence 1 mL = 1 mg Sb
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Type Standard
Quantity 500 mL
Chemical Name or Material Antimony ICP Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.