Learn More
Strem, An Ascensus Company CAS# 6923-52-0. 50g. Antimony(III) acetate, 97%. MFCD00014974
SDP
Catalog No. 5090115658
Encompass
CAS# 6923-52-0. 50g. Antimony(III) acetate, 97%. MFCD00014974. Molecular Weight: 298.88. Molecular Formula: Sb(OOCCH3)3. Color/form: off-white pwdr. Strem# 93-5111. http://www.strem.com/catalog/v/93-5111/
Catalog No. 50-901-15658 Supplier Strem, An Ascensus Company Supplier No. 93511150G
CAS# 6923-52-0. 50g. Antimony(III) acetate, 97%. MFCD00014974. Molecular Weight: 298.88. Molecular Formula: Sb(OOCCH3)3. Color/form: off-white pwdr. Strem# 93-5111. http://www.strem.com/catalog/v/93-5111/