Learn More
Strem, An Ascensus Company CAS# 10025-91-9. 250g. Antimony(III) chloride (99%-Sb). MFCD00011212
SDP
Catalog No. 5090115683
Encompass
CAS# 10025-91-9. 250g. Antimony(III) chloride (99%-Sb). MFCD00011212. Molecular Weight: 228.11. Molecular Formula: SbCl3. Color/form: off-white xtl. Strem# 93-5131. http://www.strem.com/catalog/v/93-5131/
Catalog No. 50-901-15683 Supplier Strem, An Ascensus Company Supplier No. 935131250G
CAS# 10025-91-9. 250g. Antimony(III) chloride (99%-Sb). MFCD00011212. Molecular Weight: 228.11. Molecular Formula: SbCl3. Color/form: off-white xtl. Strem# 93-5131. http://www.strem.com/catalog/v/93-5131/