Learn More
Antimony(III) fluoride, 99+%, Thermo Scientific Chemicals
Description
Antimony(III) fluoride is used as a reagent in inorganic and organofluorine chemistry. It acts as a fluorination reagent and electroplating agent. Further, it is used in the preparation of dye, pottery, porcelains, ceramic enamels and glazes. It is utilized in the preparation of Freon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 376°C |
Melting Point | 292°C |
Physical Form | Crystalline |
Quantity | 500 g |
UN Number | UN2923 |
Merck Index | 14,709 |
Solubility Information | Soluble in water,methanol and acetone. Insoluble in ammonia. |
Formula Weight | 178.75 |
Percent Purity | ≥99% |
Odor | Odorless |
Show More |
Safety and Handling
Hazard Category | H301+H311+H331-H314-H335 |
Hazard Statement | GHS H Statement H301-H311-H331 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. |
Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: II |
EINECSNumber | 232-009-2 |
RTECSNumber | CC5150000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.