Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony(III) fluoride, 99+%, Thermo Scientific Chemicals

Catalog Number AA1149222
Click to view available options
Quantity:
100 g
500 g
7783-56-4
F3Sb
178.755
MFCD00011218
GUNJVIDCYZYFGV-UHFFFAOYSA-K
antimony trifluoride, trifluorostibine, antimony fluoride, antimony iii fluoride, stibine, trifluoro, trifluoroantimony, antimoine fluorure, antimonous fluoride, antimoine fluorure french, hsdb 438
24554
trifluorostibane
F[Sb](F)F
7783-56-4
F3Sb
178.755
MFCD00011218
GUNJVIDCYZYFGV-UHFFFAOYSA-K
antimony trifluoride, trifluorostibine, antimony fluoride, antimony iii fluoride, stibine, trifluoro, trifluoroantimony, antimoine fluorure, antimonous fluoride, antimoine fluorure french, hsdb 438
24554
trifluorostibane
F[Sb](F)F

Antimony(III) fluoride is used as a reagent in inorganic and organofluorine chemistry. It acts as a fluorination reagent and electroplating agent. Further, it is used in the preparation of dye, pottery, porcelains, ceramic enamels and glazes. It is utilized in the preparation of Freon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 376°C
Melting Point 292°C
Physical Form Crystalline
Quantity 100 g
UN Number UN2923
Merck Index 14,709
Solubility Information Soluble in water,methanol and acetone. Insoluble in ammonia.
Formula Weight 178.75
Percent Purity ≥99%
Odor Odorless
Sensitivity Moisture sensitive
Density 4.379 g/mL
Chemical Name or Material Antimony(III) fluoride
Show More Show Less
Hazard Category H301+H311+H331-H314-H335
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 232-009-2
RTECSNumber CC5150000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit