Learn More
Strem, An Ascensus Company CAS# 7790-44-5. 25g. Antimony(III) iodide (99.9%-Sb). MFCD00016322
SDP
Catalog No. 5090115667
Encompass
CAS# 7790-44-5. 25g. Antimony(III) iodide (99.9%-Sb). MFCD00016322. Molecular Weight: 502.46. Molecular Formula: SbI3. Color/form: red pwdr. Strem# 93-5116. http://www.strem.com/catalog/v/93-5116/
Catalog No. 50-901-15667 Supplier Strem, An Ascensus Company Supplier No. 93511625G
CAS# 7790-44-5. 25g. Antimony(III) iodide (99.9%-Sb). MFCD00016322. Molecular Weight: 502.46. Molecular Formula: SbI3. Color/form: red pwdr. Strem# 93-5116. http://www.strem.com/catalog/v/93-5116/