Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony(III) methoxide, 99.5% (metals basis), Thermo Scientific™

Catalog No. AA4285304
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4285304 2 g
AA4285309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4285304 Supplier Thermo Scientific Chemicals Supplier No. 4285304

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29671-18-9
Molecular Formula C3H9O3Sb
Molecular Weight (g/mol) 214.862
MDL Number MFCD00014882
InChI Key KEUYHGXCOWNTEJ-UHFFFAOYSA-N
Synonym antimony iii methoxide, acmc-20alrv, antimony 3+ trimethoxide, antimony 3+ tris methoxide, antimony 3+ ion tris methoxide
PubChem CID 22289938
IUPAC Name antimony(3+);methanolate
SMILES C[O-].C[O-].C[O-].[Sb+3]

Specifications

Melting Point 124°C
Flash Point >110°C (230°F)
Quantity 2 g
UN Number UN3181
Sensitivity Air & Moisture Sensitive
Formula Weight 214.8
Percent Purity 99.5%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Antimony (III) methoxide
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
P273
Avoid release to the environment.
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.