Learn More
Antimony(III) oxide, 99.9% (metals basis)
Description
Flame retardant for wide range of plastics, rubbers, paper and textiles . Catalyst in PET production. Activator in glass industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1550°C (sublimation) |
| Melting Point | 656°C |
| Color | White |
| Physical Form | Sintered Lump |
| Quantity | 250 g |
| Odor | Odorless |
| Assay Percent Range | 99.99% (Metals basis) |
| Density | 5.67 |
| Solubility Information | Very-slightly soluble in cold water. Slightly soluble in hot water,dilute sulfuric acid,and nitric acid. Soluble in solutions of alkali hydroxide and sulfides,hydrochloric acid,tartaric acid,and acetic acid |
| Formula Weight | 291.5 |
| Show More |
Safety and Handling
| Hazard Category | H351 |
| Hazard Statement | Do not breathe dust. Wear suitable protective clothing and gloves |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| EINECSNumber | 215-175-0 |
| RTECSNumber | CC5650000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.