Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony(III) oxide, 99+%

Catalog No. p-4833913
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC213471000 100 g
AC213470010 1 kg
AC213470051 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC213471000 Supplier Thermo Scientific Chemicals Supplier No. 213471000
Only null left
Add to Cart

CAS: 1309-64-4 | O3Sb2 | 291.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-64-4
Molecular Formula O3Sb2
Molecular Weight (g/mol) 291.52
MDL Number MFCD00011214
InChI Key GHPGOEFPKIHBNM-UHFFFAOYSA-N
Synonym Antimony trioxide
IUPAC Name diantimony(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sb+3].[Sb+3]

Specifications

Boiling Point 1550°C
Melting Point 656°C
Quantity 100 g
Assay Percent Range 99% min. (Sb2O3)
Linear Formula Sb2O3
Merck Index 15,699
Solubility Information Solubility in water: 2.7mg/l (20°C). Other solubilities: slightly soluble in hot water,soluble in conc. hcl,acetic acid,sulfuric acids,,solutions of alkali hydroxides
Formula Weight 291.42
Percent Purity ≥99%
Packaging Plastic Bottle
Chemical Name or Material Antimony(III) oxide
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 215-175-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.