Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony(III) oxide, Puratronic™, 99.999% (metals basis)

Catalog No. AA1064118
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1064118 50 g
AA1064106 5 g
AA1064114 25 g
AA1064122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1064118 Supplier Thermo Scientific Chemicals Supplier No. 01064118
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1309-64-4 | O3Sb2 | 291.52 g/mol

Antimony(III) oxide finds use as flame retardant wherein it synergistically acts with halogenated materials. It is employed in opacifying glasses, ceramics and enamels. In the preparation of optical and ruby glass, antimony trioxide serves as a bubble remover. Further, it is a useful catalyst in the production of polyethylene terephthalate (PET plastic) and the vulcanization of rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-64-4
Molecular Formula O3Sb2
Molecular Weight (g/mol) 291.52
MDL Number MFCD00011214
InChI Key GHPGOEFPKIHBNM-UHFFFAOYSA-N
IUPAC Name diantimony(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sb+3].[Sb+3]

Specifications

Boiling Point 1550°C (sublimation)
Melting Point 656°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Very slightly soluble in water; soluble in potassium hydroxide,hydrochloricacid and acetic acid; insoluble in organic solvents.
Formula Weight 291.5
Odor Odorless
Density 5.67
Chemical Name or Material Antimony(III) oxide
Show More Show Less
Hazard Category H351
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 215-175-0
RTECSNumber CC5650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.