Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Antimony(III) sulfate

Catalog No. 1078350G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-783-50G 50 g
10-783-10G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-783-50G Supplier Sigma Aldrich Supplier No. 1078350G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Antimony(III) sulfate may be used as a catalyst for the imino Diels-Alder reaction of Schiff's bases with N-vinylpyrrolidin-2-one to yield 2-aryl-4-(20 -oxopyrrolidinyl-10 )-1,2,3,4- tetrahydroquinolines.

Specifications

CAS 7446-32-4
Quantity 50 g
Molecular Formula O12S3Sb2
Linear Formula Sb2(SO4)3
MDL Number MFCD00016318
Synonym Antimony trisulfate; Diantimony trisulfate
Molecular Weight (g/mol) 531.71
Percent Purity ≥95.0%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.