Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony(III) telluride, 99.999% (metals basis)

Catalog No. AA3628209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3628209 10 g
AA3628218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3628209 Supplier Thermo Scientific Chemicals Supplier No. 03628209
Add to Cart

For vacuum deposition. Activator in glass industry. Paints and Adhesives industries. Pigments, Ceramic frites. As semiconductor material, it has a large thermoelectric effect and is therefore used in solid state refrigerators. | CAS: 1327-50-0 | Sb2Te3 | 626.32 g/mol

For vacuum deposition. Activator in glass industry. Paints and Adhesives industries. Pigments, Ceramic frites. As semiconductor material, it has a large thermoelectric effect and is therefore used in solid state refrigerators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1327-50-0
Molecular Formula Sb2Te3
Molecular Weight (g/mol) 626.32
MDL Number MFCD00016320
InChI Key BPDQXJZWVBPDSN-UHFFFAOYSA-N
Synonym antimony telluride, tellanylidenestibanyl tellanyl stibanetellone, antimon iii tellurid, antimony iii telluride, antimony telluride,, antimony iii telluride, beads, antimony iii telluride, powder,-325 mesh trace metals basis
PubChem CID 6369653
IUPAC Name tellanylideneantimony;tellurium
SMILES [Sb]=[Te].[Sb]=[Te].[Te]

Specifications

Melting Point 629°C
Physical Form Lump
Quantity 10 g
UN Number UN1549
Solubility Information Insoluble in water.
Formula Weight 626.3
Percent Purity 99.999%
Packaging Plastic bottle
Specific Gravity 6.5
Density 6.5
Chemical Name or Material Antimony(III) telluride
Show More Show Less
Hazard Category H302+H332
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 215-480-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.