Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony, plasma standard solution, Specpure™ Sb 1000μg/mL

Catalog No. AA41682AP
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41682AP 500 mL
AA41682AD 50 mL
AA41682AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA41682AP Supplier Thermo Scientific Chemicals Supplier No. 041682AP
Add to Cart

missing translation for 'validMainframeMsg'

Sb in H2O/tartaric acid/tr. HNO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula Sb in H2O/tartaric acid/tr. HNO3
MDL Number MFCD00134030

Specifications

Physical Form Liquid
Concentration Sb 1000 μg/mL
Type Analytical Standard
Quantity 500 mL
Grade Specpure
Chemical Name or Material Antimony, plasma standard solution, Specpure™
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.